Assay Design
Gene Transcripts
- Gene Symbol
- Zdhhc9
- Entrez Gene ID
-
208884
- Gene Name
- zinc finger, DHHC domain containing 9
- Gene Aliases
-
6430508G22, 9530098M12Rik
- Chromosome Location
-
Chr.X: 48171967 - 48208702 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK043810.1 |
- |
2 - 3 |
400 |
135 |
View Full Product Details