Assay Design
Gene Transcripts
- Gene Symbol
- Gpatch2
- Entrez Gene ID
-
67769
- Gene Name
- G patch domain containing 2
- Gene Aliases
-
5830433G22Rik, 5830436K05Rik, AI427714, AI447508, AW107440, AW491060, Gpatc2
- Chromosome Location
-
Chr.1: 187215393 - 187369480 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK032734.1 |
- |
2 - 3 |
863 |
67 |
View Full Product Details