Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285470.1 | NP_001272399.1 | 1 - 2 | 130 | 98 |
NM_001285471.1 | NP_001272400.1 | 1 - 2 | 130 | 98 | |
GenBank mRNA | AK089927.1 | - | 1 - 2 | 104 | 98 |
AK162343.1 | - | 1 - 2 | 92 | 98 |