Assay Design



Gene Transcripts

Gene Symbol
Msh4
Entrez Gene ID
55993
Gene Name
mutS homolog 4 (E. coli)
Gene Aliases
4930485C04Rik, AV144863, mMsh4
Chromosome Location
Chr.3: 153857141 - 153906138 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282054.1 NP_001268983.1 19 - 20 2661 136
NM_031870.3 NP_114076.1 18 - 19 2752 136
GenBank mRNA AF178957.1 - 18 - 19 2456 136
AF298655.1 - 18 - 19 2752 136
AK029837.1 - 19 - 20 2661 136
AK076820.1 - 2 - 3 1658 136
AK161474.1 - 18 - 19 2502 136
AY351585.1 - 18 - 19 2706 136
AY351587.1 - 7 - 8 1703 136
AY351590.1 - 10 - 11 2168 136
AY351591.1 - 11 - 12 2192 136
BC138163.1 - 18 - 19 2643 136
BC145838.1 - 18 - 19 2643 136


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AY351588.1 - 1642 Chr. - - [-] on Build 136

View Full Product Details