Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030294.1 | NP_001025465.1 | 3 - 4 | 564 | 130 |
GenBank mRNA | AK133536.1 | - | 3 - 4 | 566 | 130 |
BC141127.1 | - | 3 - 4 | 615 | 130 | |
BC145436.1 | - | 3 - 4 | 615 | 130 |