Assay Design



Gene Transcripts

Gene Symbol
Plin1
Entrez Gene ID
103968
Gene Name
perilipin 1
Gene Aliases
6030432J05Rik, Peri, Plin
Chromosome Location
Chr.7: 79721164 - 79732904 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113471.1 NP_001106942.1 3 - 4 299 76
NM_175640.2 NP_783571.2 3 - 4 355 76
GenBank mRNA AK031445.1 - 2 - 3 280 76
AY161165.1 - 2 - 3 250 76
BC096685.1 - 3 - 4 300 76
BY723040.1 - 3 - 4 414 76

View Full Product Details