Assay Design



Gene Transcripts

Gene Symbol
Pacrg
Entrez Gene ID
69310
Gene Name
PARK2 co-regulated
Gene Aliases
1700008H23Rik
Chromosome Location
Chr.17: 10403012 - 10840191 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027032.2 NP_081308.1 1 - 2 431 79
GenBank mRNA AK005771.1 - 1 - 2 427 79
AK135510.1 - 1 - 2 433 79
BC120740.1 - 1 - 2 409 79

View Full Product Details