Assay Design



Gene Transcripts

Gene Symbol
Nop9
Entrez Gene ID
67842
Gene Name
NOP9 nucleolar protein
Gene Aliases
2610027L16Rik
Chromosome Location
Chr.14: 55745693 - 55755634 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026403.3 NP_080679.3 6 - 7 1363 81
GenBank mRNA AK011570.1 - 6 - 7 1353 81
AK015585.1 - 6 - 7 1113 81
AK032873.1 - 6 - 7 1347 81
AK037831.1 - 6 - 7 1351 81
AK050912.1 - 6 - 7 1354 81
AK083861.1 - 6 - 7 1232 81
AK139689.1 - 6 - 7 1314 81
AK146020.1 - 6 - 7 1318 81
AK146687.1 - 6 - 7 1335 81
AK152732.1 - 6 - 7 1337 81
AK154387.1 - 6 - 7 1355 81
AK159798.1 - 6 - 7 1337 81
BC018523.1 - 6 - 7 1343 81

View Full Product Details