Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012012.4 | NP_036142.2 | 13 - 14 | 2346 | 148 |
GenBank mRNA | AJ238213.1 | - | 11 - 12 | 2201 | 148 |
AK028728.1 | - | 13 - 14 | 2327 | 148 | |
AK166425.1 | - | 13 - 14 | 148 | ||
BC006671.1 | - | 13 - 14 | 2310 | 148 |