Assay Design
Gene Transcripts
- Gene Symbol
- Rpl34
- Entrez Gene ID
-
68436
- Gene Name
- ribosomal protein L34
- Gene Aliases
-
1100001I22Rik
- Chromosome Location
-
Chr.3: 130726827 - 130730398 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_026724.2 |
NP_081000.1 |
1 - 2 |
218 |
65 |
GenBank mRNA |
BC058118.1 |
- |
1 - 2 |
189 |
65 |
View Full Product Details