Assay Design



Gene Transcripts

Gene Symbol
4833412C05Rik
Entrez Gene ID
73904
Gene Name
RIKEN cDNA 4833412C05 gene
Gene Aliases
-
Chromosome Location
Chr.7: 67784538 - 67803496 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_045954.1 2 - 3 330 116
GenBank mRNA AK014683.1 - 2 - 3 331 116


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY714180.1 - 2 - 3 320 Chr. 7 - 67784775 - 67803486 [-] on Build GRCm38 116
GenBankmRNA DV069001.1 - 3 - 4 521 Chr. 7 - 67784951 - 67804118 [-] on Build GRCm38 116


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream synemin, intermediate filament protein Synm 4930412K21Rik, AI852401, Dmn, E130104F11, Syn, Synemin -25.0kb Sense Chr. - 67730161 - 67759742 on Build GRCm38
Downstream insulin-like growth factor I receptor Igf1r A330103N21Rik, CD221, D930020L01, IGF-1R, hyft 165.7kb Anti-sense Chr. - 67952257 - 68233668 on Build GRCm38

View Full Product Details