Assay Design



Gene Transcripts

Gene Symbol
Adck3
Entrez Gene ID
67426
Gene Name
aarF domain containing kinase 3
Gene Aliases
4632432J16Rik, AI462003, Cabc1, mKIAA0451
Chromosome Location
Chr.1: 180165238 - 180196020 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163290.1 NP_001156762.1 11 - 12 1453 60
NM_023341.3 NP_075830.2 11 - 12 1551 60
GenBank mRNA AJ278735.1 - 11 - 12 1431 60
AK004177.1 - 4 - 5 436 60
AK004791.1 - 11 - 12 1493 60
AK014605.2 - 11 - 12 1493 60
AK194890.1 - 2 - 3 176 60
AK199890.1 - - 176 60
AK200253.1 - 2 - 3 176 60
AK207786.1 - 2 - 3 176 60
BC030937.1 - 11 - 12 1499 60
U31629.1 - 9 - 10 1092 60

View Full Product Details