Assay Design



Gene Transcripts

Gene Symbol
Atg14
Entrez Gene ID
100504663
Gene Name
autophagy related 14
Gene Aliases
4832427M01, Atg14L, D14Ertd114e, D14Ertd436e
Chromosome Location
Chr.14: 47540893 - 47568434 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172599.4 NP_766187.1 7 - 8 1019 95
GenBank mRNA AK029967.1 - 7 - 8 1000 95
AK173037.1 - 1 - 2 73 95
BC057360.1 - 6 - 7 739 95
BC090995.1 - 7 - 8 999 95

View Full Product Details