Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033272.1 | NP_001028444.1 | 4 - 5 | 2349 | 69 |
GenBank mRNA | AK220313.1 | - | 1 - 2 | 71 | 69 |
BC035942.1 | - | 2 - 3 | 368 | 69 | |
BC138455.1 | - | 4 - 5 | 2306 | 69 | |
BC145285.1 | - | 4 - 5 | 2306 | 69 | |
CD352148.1 | - | 1 - 2 | 113 | 69 |