Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016694.3 | NP_057903.1 | 7 - 8 | 968 | 71 |
GenBank mRNA | AB019558.1 | - | 7 - 8 | 939 | 71 |
AF250293.1 | - | 7 - 8 | 968 | 71 | |
BC113204.1 | - | 7 - 8 | 951 | 71 | |
FJ555554.1 | - | 5 - 6 | 721 | 71 | |
FJ555555.1 | - | 2 - 3 | 379 | 71 | |
FJ555556.1 | - | 6 - 7 | 701 | 71 | |
FJ555557.1 | - | 6 - 7 | 826 | 71 |