Assay Design



Gene Transcripts

Gene Symbol
Nova2
Entrez Gene ID
384569
Gene Name
neuro-oncological ventral antigen 2
Gene Aliases
Gm1424
Chromosome Location
Chr.7: 18925888 - 18965319 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029877.3 NP_001025048.2 2 - 3 425 75
GenBank mRNA BF470094.1 - 1 - 2 47 75
CJ126972.1 - 2 - 3 384 75

View Full Product Details