Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301737.1 | NP_001288666.1 | 16 - 17 | 1898 | 86 |
NM_010065.3 | NP_034195.2 | 15 - 16 | 1886 | 86 | |
NR_125959.1 | 16 - 17 | 1898 | 86 | ||
GenBank mRNA | AK144142.1 | - | 15 - 16 | 1837 | 86 |
AK220483.1 | - | 16 - 17 | 1840 | 86 | |
BC034679.1 | - | 16 - 17 | 1852 | 86 | |
BC058623.1 | - | 16 - 17 | 1832 | 86 | |
BM950315.1 | - | 2 - 3 | 291 | 86 | |
CO425806.1 | - | 2 - 3 | 217 | 86 | |
L31396.1 | - | 11 - 12 | 1067 | 86 | |
L31397.1 | - | 15 - 16 | 1839 | 86 |