Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008523.2 | NP_032549.2 | 13 - 14 | 1625 | 83 |
NM_203345.2 | NP_976220.2 | 19 - 20 | 2471 | 83 | |
NM_206941.1 | NP_996824.1 | 14 - 15 | 1808 | 83 | |
NM_206942.2 | NP_996825.2 | 18 - 19 | 2288 | 83 | |
GenBank mRNA | AK157478.1 | - | 11 - 12 | 1622 | 83 |
BC047138.1 | - | 13 - 14 | 1625 | 83 | |
BC055898.1 | - | 10 - 11 | 1694 | 83 | |
BC141299.1 | - | 18 - 19 | 2264 | 83 | |
M90470.1 | - | 19 - 20 | 2471 | 83 | |
X07984.1 | - | 13 - 14 | 1599 | 83 | |
X52621.1 | - | 13 - 14 | 1614 | 83 |