Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008105.3 | NP_032131.2 | 2 - 3 | 1624 | 78 |
NM_023887.4 | NP_076376.3 | 2 - 3 | 1602 | 78 | |
NM_133219.1 | NP_573482.1 | 2 - 3 | 1264 | 78 | |
GenBank mRNA | AB037597.1 | - | 2 - 3 | 1015 | 78 |
AK054226.1 | - | 2 - 3 | 1139 | 78 | |
AK077598.1 | - | 2 - 3 | 1239 | 78 | |
AK080061.1 | - | 2 - 3 | 1191 | 78 | |
AK146638.1 | - | 2 - 3 | 366 | 78 | |
AK146679.1 | - | 2 - 3 | 366 | 78 | |
AK169167.1 | - | 2 - 3 | 366 | 78 | |
AY236873.1 | - | 2 - 3 | 1499 | 78 | |
AY236874.1 | - | 2 - 3 | 1402 | 78 | |
AY236875.1 | - | 2 - 3 | 1297 | 78 | |
AY435148.1 | - | 2 - 3 | 1015 | 78 | |
AY435149.1 | - | 2 - 3 | 1015 | 78 | |
AY435150.1 | - | 2 - 3 | 1015 | 78 | |
BC054845.1 | - | 2 - 3 | 1246 | 78 | |
BC094572.1 | - | 2 - 3 | 1602 | 78 | |
BQ892107.1 | - | 2 - 3 | 773 | 78 |