Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190179.1 | NP_001177108.1 | 2 - 3 | 311 | 138 |
NM_027091.4 | NP_081367.1 | 2 - 3 | 291 | 138 | |
GenBank mRNA | AF411517.1 | - | 2 - 3 | 270 | 138 |
AK009187.1 | - | 2 - 3 | 239 | 138 | |
AK089997.1 | - | 2 - 3 | 290 | 138 | |
AK136109.1 | - | 2 - 3 | 254 | 138 | |
AK145775.1 | - | 2 - 3 | 238 | 138 | |
BC048814.1 | - | 2 - 3 | 246 | 138 | |
BY272877.1 | - | 2 - 3 | 291 | 138 | |
CA560987.1 | - | 2 - 3 | 311 | 138 | |
DV653588.1 | - | 2 - 3 | 286 | 138 |