Assay Design



Gene Transcripts

Gene Symbol
Per1
Entrez Gene ID
18626
Gene Name
period circadian clock 1
Gene Aliases
Per, m-rigui, mPer1
Chromosome Location
Chr.11: 69095140 - 69109965 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159367.1 NP_001152839.1 17 - 18 2386 62
NM_011065.4 NP_035195.2 17 - 18 2401 62
GenBank mRNA AB002108.1 - 16 - 17 2215 62
AF022992.1 - 17 - 18 2440 62
AK081813.1 - 17 - 18 2399 62
AK148202.1 - 17 - 18 2393 62
AK172958.1 - 17 - 18 2386 62
BC039768.1 - 16 - 17 2286 62
BC091645.1 - 17 - 18 2388 62

View Full Product Details