Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033294.3 | NP_001028466.2 | 8 - 9 | 1060 | 69 |
GenBank mRNA | AK030888.1 | - | 8 - 9 | 1007 | 69 |
AK144739.1 | - | 8 - 9 | 773 | 69 | |
AK166327.1 | - | 8 - 9 | 730 | 69 | |
BC066017.1 | - | 8 - 9 | 697 | 69 | |
BC132351.1 | - | 8 - 9 | 750 | 69 |