Assay Design



Gene Transcripts

Gene Symbol
Ntng2
Entrez Gene ID
171171
Gene Name
netrin G2
Gene Aliases
2610016D08Rik, Lmnt2
Chromosome Location
Chr.2: 29194821 - 29248040 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB052337.1 - 6 - 7 1231 59
AB052338.1 - 5 - 6 1156 59
AF475080.1 - 6 - 7 1231 59

View Full Product Details