Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021516.4 | NP_067491.2 | 1 - 2 | 594 | 58 |
NM_022801.4 | NP_073712.2 | 1 - 2 | 594 | 58 | |
GenBank mRNA | AF240782.1 | - | 1 - 2 | 139 | 58 |
AF240783.1 | - | 1 - 2 | 139 | 58 | |
AI850379.1 | - | 1 - 2 | 132 | 58 | |
AK083683.1 | - | 1 - 2 | 595 | 58 | |
AK132247.1 | - | 1 - 2 | 578 | 58 | |
AK166157.1 | - | 1 - 2 | 583 | 58 | |
BC066071.1 | - | 1 - 2 | 568 | 58 |