Assay Design



Gene Transcripts

Gene Symbol
Skap2
Entrez Gene ID
54353
Gene Name
src family associated phosphoprotein 2
Gene Aliases
2610021A10Rik, AA960083, BB137539, RA70, SKAP-HOM, Saps, Scap2, mSKAP55R
Chromosome Location
Chr.6: 51859165 - 52012549 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018773.2 NP_061243.1 12 - 13 1236 75
GenBank mRNA AB014485.1 - 12 - 13 1153 75
AF051324.1 - 12 - 13 1151 75
AK076000.1 - 12 - 13 1181 75
AK144289.1 - 12 - 13 1173 75
BC003711.1 - 12 - 13 1236 75

View Full Product Details