Assay Design



Gene Transcripts

Gene Symbol
Scrn1
Entrez Gene ID
69938
Gene Name
secernin 1
Gene Aliases
2810019K23Rik, 6330535A03Rik, AI852905, SES1, mKIAA0193
Chromosome Location
Chr.6: 54472300 - 54566499 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027268.2 NP_081544.1 3 - 4 481 65
GenBank mRNA AK012765.1 - 3 - 4 509 65
AK032573.1 - 3 - 4 508 65
AK050481.1 - 3 - 4 499 65
AK082953.1 - 3 - 4 481 65
AK089968.1 - 3 - 4 471 65
AK129084.1 - 3 - 4 468 65
AK145904.1 - 3 - 4 483 65
BC023889.1 - 3 - 4 468 65
EF410099.1 - - 432 65

View Full Product Details