Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027268.2 | NP_081544.1 | 7 - 8 | 1226 | 67 |
GenBank mRNA | AK012765.1 | - | 7 - 8 | 1254 | 67 |
AK032573.1 | - | 7 - 8 | 1253 | 67 | |
AK050481.1 | - | 7 - 8 | 1244 | 67 | |
AK082953.1 | - | 7 - 8 | 1226 | 67 | |
AK089968.1 | - | 7 - 8 | 1217 | 67 | |
AK129084.1 | - | 7 - 8 | 1213 | 67 | |
AK145904.1 | - | 7 - 8 | 1228 | 67 | |
BC023889.1 | - | 7 - 8 | 1213 | 67 | |
BC058619.1 | - | 1 - 2 | 2765 | 67 |