Assay Design



Gene Transcripts

Gene Symbol
Surf6
Entrez Gene ID
20935
Gene Name
surfeit gene 6
Gene Aliases
D2Wsu129e, Surf-6
Chromosome Location
Chr.2: 26890772 - 26902813 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009298.3 NP_033324.1 2 - 3 328 105
GenBank mRNA AK077989.1 - 2 - 3 330 105
AK132194.1 - 2 - 3 364 105
BC037634.1 - 2 - 3 375 105
X92842.1 - 2 - 3 311 105

View Full Product Details