Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207234.2 | NP_997117.2 | 7 - 8 | 1246 | 121 |
GenBank mRNA | AB221675.1 | - | 7 - 8 | 1178 | 121 |
AK133982.1 | - | 7 - 8 | 1166 | 121 | |
BC060147.1 | - | 7 - 8 | 1172 | 121 |