Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019539.3 | NP_062412.1 | 5 - 6 | 841 | 64 |
GenBank mRNA | AF250837.1 | - | 4 - 5 | 628 | 64 |
AY014779.1 | - | 5 - 6 | 701 | 64 | |
BC050198.1 | - | 4 - 5 | 937 | 64 | |
BC064740.1 | - | 5 - 6 | 841 | 64 |