Assay Design



Gene Transcripts

Gene Symbol
Fam73b
Entrez Gene ID
108958
Gene Name
family with sequence similarity 73, member B
Gene Aliases
5730472N09Rik, AA408683, AI790341, C9orf54, R74766
Chromosome Location
Chr.2: 30364198 - 30385520 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242407.1 NP_001229336.1 1 - 2 70 55
GenBank mRNA AK040713.1 - 1 - 2 39 55
AK077618.1 - 1 - 2 31 55
AK140122.1 - 1 - 2 56 55
AK154200.1 - 1 - 2 59 55
AK165996.1 - 1 - 2 61 55
BY151776.1 - 1 - 2 71 55

View Full Product Details