Assay Design



Gene Transcripts

Gene Symbol
Fam73b
Entrez Gene ID
108958
Gene Name
family with sequence similarity 73, member B
Gene Aliases
5730472N09Rik, AA408683, AI790341, C9orf54, R74766
Chromosome Location
Chr.2: 30364198 - 30385520 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242407.1 NP_001229336.1 7 - 8 1007 64
NM_175392.3 NP_780601.1 7 - 8 1175 64
GenBank mRNA AK077618.1 - 7 - 8 968 64
AK131152.1 - 4 - 5 1857 64
AK140122.1 - 7 - 8 993 64
AK154200.1 - 7 - 8 996 64
AK165996.1 - 7 - 8 999 64
BC051404.1 - 7 - 8 1175 64

View Full Product Details