Assay Design



Gene Transcripts

Gene Symbol
Fam73b
Entrez Gene ID
108958
Gene Name
family with sequence similarity 73, member B
Gene Aliases
5730472N09Rik, AA408683, AI790341, C9orf54, R74766
Chromosome Location
Chr.2: 30364198 - 30385520 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242407.1 NP_001229336.1 8 - 9 1102 70
NM_175392.3 NP_780601.1 8 - 9 1270 70
GenBank mRNA AK077618.1 - 8 - 9 1063 70
AK131152.1 - 5 - 6 1952 70
AK140122.1 - 8 - 9 1088 70
AK154200.1 - 8 - 9 1091 70
AK165996.1 - 8 - 9 1094 70
BC051404.1 - 8 - 9 1270 70

View Full Product Details