Assay Design



Gene Transcripts

Gene Symbol
Pde11a
Entrez Gene ID
241489
Gene Name
phosphodiesterase 11A
Gene Aliases
6330414F14Rik, A630086N24Rik, Gm350, PDE11A1
Chromosome Location
Chr.2: 75987026 - 76339145 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081033.1 NP_001074502.1 19 - 20 2693 74
GenBank mRNA BC158125.1 - 19 - 20 2833 74

View Full Product Details