Assay Design



Gene Transcripts

Gene Symbol
Pde11a
Entrez Gene ID
241489
Gene Name
phosphodiesterase 11A
Gene Aliases
6330414F14Rik, A630086N24Rik, Gm350, PDE11A1
Chromosome Location
Chr.2: 75987026 - 76339145 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081033.1 NP_001074502.1 7 - 8 1618 65
GenBank mRNA AK042383.1 - 5 - 6 489 65
BC158125.1 - 7 - 8 1758 65

View Full Product Details