Assay Design



Gene Transcripts

Gene Symbol
Dgkz
Entrez Gene ID
104418
Gene Name
diacylglycerol kinase zeta
Gene Aliases
E130307B02Rik, ENSMUSG00000075035, F730209L11Rik, mDGK[z]
Chromosome Location
Chr.2: 91932822 - 91976443 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166597.1 NP_001160069.1 8 - 9 1363 66
NM_138306.2 NP_612179.2 7 - 8 762 66
GenBank mRNA AK087503.1 - 8 - 9 1364 66
AK170936.1 - 4 - 5 385 66
AK210058.1 - 3 - 4 174 66
BC016217.1 - 7 - 8 1709 66
BC049228.1 - 7 - 8 798 66

View Full Product Details