Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276359.1 | NP_001263288.1 | 12 - 13 | 4250 | 60 |
NM_001276360.1 | NP_001263289.1 | 13 - 14 | 4283 | 60 | |
NM_001276361.1 | NP_001263290.1 | 13 - 14 | 4310 | 60 | |
GenBank mRNA | AK035221.1 | - | 5 - 6 | 2762 | 60 |