Assay Design



Gene Transcripts

Gene Symbol
Camsap1
Entrez Gene ID
227634
Gene Name
calmodulin regulated spectrin-associated protein 1
Gene Aliases
9530003A05Rik, C77823, PRO2405
Chromosome Location
Chr.2: 25926838 - 25983452 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276359.1 NP_001263288.1 2 - 3 767 62
NM_001276360.1 NP_001263289.1 2 - 3 767 62
NM_001276361.1 NP_001263290.1 2 - 3 767 62
GenBank mRNA AK137097.1 - 2 - 3 768 62
BC024715.2 - 2 - 3 482 62
BC054553.1 - 2 - 3 546 62
CD351317.1 - 1 - 2 215 62

View Full Product Details