Assay Design
Gene Transcripts
- Gene Symbol
- Spc25
- Entrez Gene ID
-
66442
- Gene Name
- SPC25, NDC80 kinetochore complex component, homolog (S. cerevisiae)
- Gene Aliases
-
2600017H08Rik, 2610205L13Rik, Spbc25
- Chromosome Location
-
Chr.2: 69193895 - 69206215 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK151554.1 |
- |
1 - 2 |
100 |
98 |
BC027121.1 |
- |
1 - 2 |
67 |
98 |
CX222514.1 |
- |
1 - 2 |
126 |
98 |
View Full Product Details