Assay Design



Gene Transcripts

Gene Symbol
Klhl23
Entrez Gene ID
277396
Gene Name
kelch-like 23
Gene Aliases
C130068N17Rik
Chromosome Location
Chr.2: 69821920 - 69836651 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177784.4 NP_808452.2 2 - 3 1486 88
GenBank mRNA AK048518.1 - 2 - 3 1385 88
BC072626.1 - 2 - 3 1486 88

View Full Product Details