Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025922.2 | NP_080198.2 | 3 - 4 | 219 | 91 |
GenBank mRNA | AB100501.1 | - | 3 - 4 | 231 | 91 |
AK009248.1 | - | 3 - 4 | 232 | 91 | |
AK138101.1 | - | 3 - 4 | 259 | 91 | |
AK139770.1 | - | 3 - 4 | 217 | 91 | |
AK153806.1 | - | 3 - 4 | 259 | 91 | |
AK153904.1 | - | 3 - 4 | 238 | 91 | |
BC026508.1 | - | 3 - 4 | 219 | 91 | |
BC094466.1 | - | 3 - 4 | 207 | 91 |