Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026580.4 | NP_080856.1 | 1 - 2 | 194 | 109 |
GenBank mRNA | AK006054.1 | - | 1 - 2 | 116 | 109 |
AK006346.1 | - | 1 - 2 | 146 | 109 | |
AK006469.1 | - | 1 - 2 | 143 | 109 | |
AK019830.1 | - | 1 - 2 | 171 | 109 | |
AK160505.1 | - | 1 - 2 | 173 | 109 | |
BC049551.1 | - | 1 - 2 | 177 | 109 | |
BC052409.1 | - | 1 - 2 | 109 | 109 |