Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019488.4 | NP_062361.1 | 2 - 3 | 262 | 84 |
GenBank mRNA | AF232061.2 | - | 2 - 3 | 219 | 84 |
AJ245936.1 | - | 2 - 3 | 239 | 84 | |
AK081806.1 | - | 2 - 3 | 279 | 84 | |
AK155792.1 | - | 2 - 3 | 290 | 84 | |
AK170319.1 | - | 2 - 3 | 275 | 84 | |
AY856043.1 | - | 2 - 3 | 223 | 84 | |
AY856044.1 | - | 2 - 3 | 223 | 84 | |
AY856045.1 | - | 2 - 3 | 223 | 84 | |
BC090993.1 | - | 2 - 3 | 262 | 84 | |
Y17802.1 | - | 2 - 3 | 244 | 84 |