Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019488.4 | NP_062361.1 | 6 - 7 | 909 | 96 |
GenBank mRNA | AF232061.2 | - | 6 - 7 | 866 | 96 |
AJ245936.1 | - | 6 - 7 | 886 | 96 | |
AK081806.1 | - | 6 - 7 | 926 | 96 | |
AK155792.1 | - | 6 - 7 | 96 | ||
AK170319.1 | - | 6 - 7 | 922 | 96 | |
AY856043.1 | - | 6 - 7 | 870 | 96 | |
BC021758.1 | - | 2 - 3 | 235 | 96 | |
BC090993.1 | - | 6 - 7 | 909 | 96 | |
Y17802.1 | - | 6 - 7 | 891 | 96 |