Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029843.1 | NP_001025014.1 | 10 - 11 | 991 | 91 |
NM_001029844.1 | NP_001025015.1 | 10 - 11 | 991 | 91 | |
NM_011705.3 | NP_035835.1 | 10 - 11 | 991 | 91 | |
GenBank mRNA | AF080252.1 | - | 9 - 10 | 904 | 91 |
AF080253.1 | - | 10 - 11 | 945 | 91 | |
AK032344.1 | - | 10 - 11 | 962 | 91 | |
AK039554.1 | - | 4 - 5 | 363 | 91 | |
AK088230.1 | - | 10 - 11 | 915 | 91 | |
AK088674.1 | - | 10 - 11 | 974 | 91 | |
AK136352.1 | - | 10 - 11 | 965 | 91 | |
AK166676.1 | - | 10 - 11 | 959 | 91 | |
BC016676.1 | - | 10 - 11 | 1026 | 91 | |
BC051072.1 | - | 10 - 11 | 971 | 91 |