Assay Design



Gene Transcripts

Gene Symbol
Vrk1
Entrez Gene ID
22367
Gene Name
vaccinia related kinase 1
Gene Aliases
51PK
Chromosome Location
Chr.12: 106010163 - 106077421 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029843.1 NP_001025014.1 5 - 6 479 149
NM_001029844.1 NP_001025015.1 5 - 6 479 149
NM_011705.3 NP_035835.1 5 - 6 479 149
GenBank mRNA AF080252.1 - 4 - 5 392 149
AF080253.1 - 5 - 6 433 149
AK032344.1 - 5 - 6 450 149
AK088674.1 - 5 - 6 462 149
AK136352.1 - 5 - 6 453 149
AK166676.1 - 5 - 6 447 149
BC016676.1 - 5 - 6 514 149
BC051072.1 - 5 - 6 459 149

View Full Product Details