Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029843.1 | NP_001025014.1 | 8 - 9 | 809 | 81 |
NM_001029844.1 | NP_001025015.1 | 8 - 9 | 809 | 81 | |
NM_011705.3 | NP_035835.1 | 8 - 9 | 809 | 81 | |
GenBank mRNA | AF080252.1 | - | 7 - 8 | 722 | 81 |
AF080253.1 | - | 8 - 9 | 763 | 81 | |
AK032344.1 | - | 8 - 9 | 780 | 81 | |
AK039554.1 | - | 2 - 3 | 181 | 81 | |
AK088230.1 | - | 8 - 9 | 733 | 81 | |
AK088674.1 | - | 8 - 9 | 792 | 81 | |
AK136352.1 | - | 8 - 9 | 783 | 81 | |
AK166676.1 | - | 8 - 9 | 777 | 81 | |
BC016676.1 | - | 8 - 9 | 844 | 81 | |
BC051072.1 | - | 8 - 9 | 789 | 81 |