Assay Design
Gene Transcripts
- Gene Symbol
- Ahnak2
- Entrez Gene ID
-
100041194
- Gene Name
- AHNAK nucleoprotein 2
- Gene Aliases
-
AI450948, Gm1185, Gm72
- Chromosome Location
-
Chr.12: 112772906 - 112802660 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK138503.1 |
- |
4 - 5 |
441 |
64 |
View Full Product Details