Assay Design
Gene Transcripts
- Gene Symbol
- Akr1c20
- Entrez Gene ID
-
116852
- Gene Name
- aldo-keto reductase family 1, member C20
- Gene Aliases
-
2610528B18Rik
- Chromosome Location
-
Chr.13: 4486972 - 4523354 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB097220.1 |
- |
8 - 9 |
939 |
113 |
BC021607.1 |
- |
8 - 9 |
947 |
113 |
View Full Product Details