Assay Design



Gene Transcripts

Gene Symbol
Chrdl1
Entrez Gene ID
83453
Gene Name
chordin-like 1
Gene Aliases
CHL, CHL1, Nrln1, VOPT
Chromosome Location
Chr.X: 143285674 - 143394262 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114385.1 NP_001107857.1 10 - 11 1647 91
GenBank mRNA AF321853.1 - 9 - 10 1123 91
BC050818.1 - 9 - 10 1137 91

View Full Product Details